1x1 image pixel for data collection
Dextran Pyrogen Free Clinical MP Biomedicals
Dextran Pyrogen Free Clinical MP Biomedicals
Dextran Pyrogen Free Clinical MP Biomedicals
Dextran Pyrogen Free Clinical MP Biomedicals
MP Biomedicals
Dextran Pyrogen Free Clinical MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB021015145050 g
Catalog number MPB0210151450
Price (SGD)
165.00
EA
Estimated availability date 21-Sep-2025
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (SGD)
165.00
EA
Add to cart
Specifications
Chemical Name or MaterialDextran
Name NotePyrogen Free
CAS9004-54-0
Molecular Formula(C6H9O5)n(C6H10O5)m
MDL NumberMFCD00130935
View more
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link