Check Order Status
Dextran, MP Biomedicals™
Element Search
Structure Search
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
MP Biomedicals
Dextran, MP Biomedicals™
Have Questions?
Contact Us
Catalog Number
Quantity
MPB0210150510
10 g
Catalog number
MPB0210150510
Price (SGD)
88.00
EA
Estimated availability date
21-Sep-2025
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (SGD)
88.00
EA
Add to cart
Specifications
Chemical Name or Material
Dextran
CAS
9004-54-0
Recommended Storage
Store at Room Temperature (15° to 30°C)
Molecular Formula
(C6H9O5)n(C6H10O5)m
MDL Number
MFCD00130935
View more
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link