▸
Organic compounds
▸
Unclassified Organic Compounds
▸
Beta-Nicotinamide Adenine Dinucleotide Reduced Disodium Salt, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Beta-Nicotinamide Adenine Dinucleotide Reduced Disodium Salt, MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0210116810
10 g
Catalog number
MPB0210116810
Price (SGD)
1,799.00
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (SGD)
1,799.00
Each
Add to cart
Specifications
Chemical Name or Material
Beta-Nicotinamide Adenine Dinucleotide Reduced Disodium Salt
CAS
606-68-8
Molecular Formula
C21H27N7Na2O14P2
MDL Number
MFCD00036200
Quantity
10 g
View more