Check Order Status
bis-Tris, ≥99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
bis-Tris, ≥99%, MP Biomedicals™
Exhibits a very small shift in dissociation constant with respect to temperature
Have Questions?
Catalog Number
Quantity
MPB0210103891
1 kg
Catalog number
MPB0210103891
Price (SGD)
1,415.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
1,415.00
EA
Add to cart
Specifications
Chemical Name or Material
bis-Tris
CAS
6976-37-0
Color
White
Physical Form
Crystalline Powder
Quantity
1 kg
View more
Zwitterionic buffer
Buffering agent
White Powder