Product Image
MP Biomedicals
β-D-Glucose, MP Biomedicals, LLC
β-D-Glucose is ≥80% pure and is used for many cell biology applications.
Have Questions?
Catalog NumberQuantity
MPB021009531010 g
Catalog number MPB0210095310
Price (SGD)
101.00
Each
Quantity:
10 g
Request bulk or custom format
Price (SGD)
101.00
Each
Specifications
Melting Point148°C to 155°C (literature)
ColorWhite
Density1.544 g/mL
Boiling Point∼410.8°C (760 mmHg (Predicted) (literature))
PackagingPlastic
View more
  • β-D-Glucose is a main source of energy for living organisms
  • Combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen
  • β-D-glucose may contain up to 10% of α-isomer