Papain from Papaya Latex
Papain from Papaya Latex
Actual product may vary
MP Biomedicals
Papain from Papaya Latex
Sulfhydryl protease from Carica papaya latex
Have Questions?
Catalog NumberQuantity
MPB0210092180100 mg
Catalog number MPB0210092180
Price (SGD)
335.00
EA
Add to cart
Quantity:
100 mg
Price (SGD)
335.00
EA
Add to cart
Papain cleaves peptide bonds of basic amino acids, leucine, or glycine. It is also hydrolyzes esters and amides.
  • CAS Number: 9001-73-4
  • EC Number: 232-627-2
  • MDL Number: MFCD00131791
  • Format: Milky white solution
  • Specific Activity: ≤10 units/mg Protein
  • Extinction coefficient: 25.0
  • Isoelectric point (pI): pH 9.6
  • Optimum pH: 6.0 to 7.0
Used to produce Fab fragments of antibodies, cell dissociation, morphological isolation of intact cortical neurons from postnatal rats. Limited papaindigestion is used for structural studies of enzymes and other proteins. In addition, papain is used in red cell serology to modify red cell surface to enhance or destroy reactivity of many red cell antigens. Useful for platelet serology studies and has also been used in enzymatic synthesis of amino acids, peptides, and other molecules.
Specifications
Concentration2X
ColorMilky White
pH6 to 7
For Use With (Application)Produce Fab fragments of Antibodies, cell dissociation, morphological isolation of intact cortical neurons from Postnatal Rats
SourcePapaya Latex
Quantity100 mg
Product TypePapain
Molecular Weight (g/mol)23000
InhibitorsSulfhydryl groups including heavy Metals, Carbonyl reagents, Aldehydes, Benzoylamidoacetonitrile, AEBSF, antipain, cystatin, E-64, leupeptin, PMSF, TLCK and TPCK
FormSolution
Unit SizeEA
Contents & Storage
4°C