Adenosine 5'-monophosphate monohydrate, 99.7%, MP Biomedicals™
Adenosine 5'-monophosphate monohydrate, 99.7%, MP Biomedicals™
Adenosine 5'-monophosphate monohydrate, 99.7%, MP Biomedicals™
MP Biomedicals
Adenosine 5'-monophosphate monohydrate, 99.7%, MP Biomedicals™
Used as a monomer in RNA. It is an ester of phosphoric acid and the nucleoside adenosine. AMP consists of a phosphate group, the sugar ribose, and the nucleobase adenine
Have Questions?
Catalog NumberQuantity
MPB02100080011 g
Catalog number MPB0210008001
Price (SGD)
69.00
Each
Quantity:
1 g
Request bulk or custom format
Price (SGD)
69.00
Each
Specifications
Chemical Name or MaterialAdenosine 5'-monophosphate monohydrate
CAS18422-05-4
ColorWhite
Assay Percent Range99.70%
Molecular FormulaC10H16N5O8P
View more