▸
Salts and Inorganics
▸
Inorganic Salts
▸
Metalloid Salts
▸
Silica, 32-63μm, 60Å, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Silica, 32-63μm, 60Å, MP Biomedicals™
For Chromatography; Chemical Synthesis
Have Questions?
Catalog Number
Quantity
MPB0202825
1 kg
Catalog number
MPB0202825
Price (SGD)
379.00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
379.00
Each
Add to cart
Specifications
Chemical Name or Material
Silica
CAS
7631-86-9
Color
White to Gray
Recommended Storage
Store at Room Temperature.
Physical Form
Powder
View more