Duraprene™ CP Sterile Neoprene Cleanroom Gloves
Duraprene™ CP Sterile Neoprene Cleanroom Gloves
Actual product may vary
Duraprene™ CP Sterile Neoprene Cleanroom Gloves
Duraprene™ CP Sterile Neoprene Cleanroom Gloves
Duraprene™ CP Sterile Neoprene Cleanroom Gloves
Sterile neoprene gloves meet all relevant ASTM standards and are recommended for ISO 5/Class 100 or higher cleanrooms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
MLL2Y72PN808
MLL2Y72PN606
MLL2Y72PN656.5
MLL2Y72PN707
MLL2Y72PN757.5
MLL2Y72PN909
Catalog number MLL2Y72PN80
Price (SGD)
496.00
CS
Add to cart
Size:
8
Price (SGD)
496.00
CS
Add to cart
  • Sterile, powder free, synthetic neoprene
  • Recommended for ISO 5/Class 100 or higher
  • Length 12 inches
  • Clean-processed and chlorinated
  • Exclusive hand molds with independent thumb design allow for natural fit and movement
Specifications
Certifications/ComplianceASTM D3577
Cleanroom ClassClass 100 (ISO 5)
AQL1
ColorNatural
Exterior FinishNo Texture
Finger Thickness (English)8 mil
Finger Thickness (Metric)0.20 mm
HandSpecific
Latex FreeYes
MaterialNeoprene
Palm Thickness (English)7.9 mil
Palm Thickness (Metric)0.20 mm
PowderedPowder Free
Size8
SterilitySterile
Thickness (English)8 mil
Thickness (Metric)0.20 mm
Manufacturing/QMS Audit StandardsISO 13485
Model2Y72PN80
SeriesDuraprene™ CP
Tensile Strength22 MPa
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCS