Flange Wedges
Actual product may vary
Flange Wedges
Flange Wedges
Micronova
Flange Wedges
Quality flange wedges for a safe workplace
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)Weight (English)Width (English)
MICW-98 in.1.66 lb.2 in.
MICW-108.5 in.2.96 lb.2 in.
Catalog number MICW-9
Price (SGD)
564.00
Each
Length (English):
8 in.
Weight (English):
1.66 lb.
Width (English):
2 in.
Price (SGD)
564.00
Each
  • Non-sparking
  • Non-magnetic
  • Corrosion-resistant
Specifications
Product TypeFlange Wedge
Blade MaterialAluminum Bronze
Length (English)8 in.
Length (Metric)20 cm
MaterialAluminum Bronze
Dimensions (L x W)8 x 2 in. (20 x 5 cm)
For Use With (Application)Non-sparking, non-magnetic, corrosion-resistant applications
Weight (English)1.66 lb.
Weight (Metric)0.7 kg
Width (English)2 in.
Width (Metric)5 cm
Unit SizeEach