T-Mop™ Covers
Actual product may vary
Micronova
T-Mop™ Covers
A range of T-mop covers with snaps that fit onto the 180° swivel TMop™ Adapter to effortlessly clean and sanitize cleanroom surfaces, including soft walls, awkward corners, tight spaces and under equipment.​ Available in a variety of fabrics to meet specific cleaning needs.
Have Questions?
Catalog NumberMaterialDimensionsColorQuantity
MICTAA12-18Microfiber Polyester, Nylon, Urethane Foam, Urethane Foam45.7 x 6.4 cm (18 x 2.5 in.)Blue12/Cs.
Catalog number MICTAA12-18
Price (SGD)
621.00
Case
Material:
Microfiber Polyester, Nylon, Urethane Foam, Urethane Foam
Dimensions:
45.7 x 6.4 cm (18 x 2.5 in.)
Color:
Blue
Quantity:
12/Cs.
Price (SGD)
621.00
Case
The Micronova TMop™ Cleanroom Mop Covers are rounded, covered sponges with snaps that fit over the 180° swivel TMop™ Adapter to easily roll down walls and soft walls as well as clean awkward corners, tight spaces and under equipment. The mops are available in a range of fabric options with an optional thicker foam layer to hold more disinfectant and facilitate wringing.
  • T-Mop covers with snap fasteners in a range of fabrics
  • Rounded covered sponges easily roll down walls and soft walls
  • Excellent on all room surfaces and equipment exteriors
  • Easily fits in awkward corners, tight spaces and under equipment
  • Fabric options include:
    • NovaPoly — 100% smooth knit polyester provides excellent disinfectant dispersal
    • PolySorb — 100% textured polyester for removing residues and dispersing disinfectant
    • MegaTex — 100% non-woven polyamide is absorbent with a mildly abrasive surface
    • Microfiber — 80% polyester, 20% polyamide is highly absorbent and effective wet or dry
    • PolyMesh — 100% polyester with a layer of mesh provides a mild scrubbing surface
  • For use with the 180° swivel TMop™ Adapter
  • Available in 10, 14 and 18-inch lengths
  • TMC6-14 - elastic single-ply slipcover, low cost option for smaller areas
  • Holds up to strong disinfectants
  • Autoclavable
Does not include adaptor and handle
Specifications
Dimensions45.7 x 6.4 cm (18 x 2.5 in.)
Product TypeMop Cover
ColorBlue
Length (English)18 in.
SterilityNon-sterile
Length (Metric)45.7 cm
AutoclavableAutoclavable
Width (Metric)6.4 cm
Width (English)2.5 in.
Resistant ToChemicals
Size45.72 cm (18 in.)
DisposableReusable
Quantity12/Cs.
Care InstructionsItem may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
IncludesThick Foam
For Use With (Application)Cleaning and Sanitization of Cleanroom Surfaces
Color-codedNo
MaterialMicrofiber Polyester, Nylon, Urethane Foam, Urethane Foam
Unit SizeCase