Specifications
Product TypeMop Cover
ColorWhite
Length (English)10 in.
SterilityNon-sterile
Length (Metric)25.4 cm
AutoclavableAutoclavable
Width (Metric)5.1 cm
Width (English)2 in.
Resistant ToChemicals
Size10 in. (Width)
DisposableReusable
Quantity24/Cs.
Care InstructionsItem may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
For Use With (Equipment)TMop Round or Square (QDT, QPT, QDST, QPST)
DescriptionTMop, PolySorb, No Sponge, 10 in.
For Use With (Application)Cleaning and Sanitization of Cleanroom Surfaces
Certifications/ComplianceCertificate of Conformance
Color-codedNo
MaterialPolyester
Unit SizeEach