Search Fisher Scientific
| Catalog Number | Product Type | Material | Length (English) |
|---|---|---|---|
| MICQDNS-22 | Squeegee | Electro-polished Stainless Steel (304) | 22 in. |
The Micronova Cleanroom NovaSqueegee™ is a stainless steel tool that comes standard with a replaceable rubber blade and features a Quick Disconnect handle attachment for optional use with the SSU series handles.