Accordion Over-Under™ Handle
Actual product may vary
Accordion Over-Under™ Handle
Accordion Over-Under™ Handle
Micronova
Accordion Over-Under™ Handle
Designed to deal with the unique space limitations in isolators and containment hoods
Have Questions?
Catalog NumberMaterial
MICQDAH-08Electropolished Stainless Steel (304), Polyethylene
Catalog number MICQDAH-08
Price (SGD)
377.00
Each
Material:
Electropolished Stainless Steel (304), Polyethylene
Price (SGD)
377.00
Each
  • Each section features a polypropylene hinge and quick disconnect to allow for attachment of additional sections and ultimately the mop adapter
  • The unique design allows the operator to fit the collapsed handle into the pass thru and once inside the isolator provides the extended reach necessary to cover all surfaces
  • Outside of the Isolator, when connected to a standard handle, the Accordion Handle allows the operator to angle the mop head to effectively and comfortably clean ‘over and under’ obstacles in a cleanroom or controlled environment
  • Autoclavable
Specifications
SterilitySterile
Width (English)8 in.
DescriptionStainless steel handle with locking swivel action that holds various adapters and works as an extension to reach over and under equipment
MaterialElectropolished Stainless Steel (304), Polyethylene
Unit SizeEach