PharmaPly™ Mop Head
Actual product may vary
Micronova
PharmaPly™ Mop Head
Ideal for covering large surface areas quickly and efficiently.
Have Questions?
Catalog number MICPPM4-MM15
Price (SGD)
819.00
Case
Price (SGD)
819.00
Case
  • Polyester/rayon blend mop head for cleaning/disinfecting pharmaceutical support areas, hospitals and laboratories
  • Designed for use with MicroMop™ adapter and handle (sold separately)
  • 15 in. long (38cm)
Specifications
Color CodeWhite
Product TypeCleanroom Mop
Length (English)15 in.
SterilitySterile
AutoclavableAutoclavable
Width (English)6 in.
DisposableReusable
For Use With (Application)Used for Covering Large Surface Areas Quickly and Efficiently, Floor
Color-codedNo
MaterialPolyester, Rayon
Unit SizeCase