PCX™ Polyethylene Tapes
Actual product may vary
Micronova
PCX™ Polyethylene Tapes
A low-density, low-outgassing polyethylene tape that is ideal for sealing, color coding, labeling, batching and food processing applications. Available in a variety of vibrant color options.
Have Questions?
Catalog NumberColorWidth (Metric)
MICPCX-34TRTransparent19.05 mm
Catalog number MICPCX-34TR
Price (SGD)
-
Color:
Transparent
Width (Metric):
19.05 mm
The Micronova PCX™ Polyethylene Tape is a low density LDPE tape, with pressure sensitive acrylic adhesive, that is low in halogens and leachable chlorides to lessen the chance of outgassing. Available in seven vibrant colors and 1 to 3-inch widths for a variety of cleanroom and food processing applications.
  • Low-density, low-outgassing polyethylene tape with
  • Pressure-sensitive acrylic adhesive
  • Ideal for sealing, color coding, labeling, batching, etc.
  • Available in dark green, red, transparent, white, yellow, black and orange
  • Excellent chemical resistance and good conformability
  • Processed and packaged in a cleanroom
  • Wound on a 3 in. plastic core
Specifications
AutoclavableNot Autoclavable
Adhesive TypeAcrylic
CoatingPolyethylene
ColorTransparent
FinishMedium Gloss
MaterialPolyethylene
Product TypeCleanroom Tape
SterilityNon-sterile
For Use With (Application)Sealing, Color Coding, Labeling, Batching and Food Processing Applications
Dimensions (L x W)33 m x 19.05 mm (36 yd. x 0.75 in.)
Length (English)36 yd.
Length (Metric)33 m
Width (English)0.75 in.
Width (Metric)19.05 mm
Unit SizeCase