Tape Dispensers
Actual product may vary
Micronova
Tape Dispensers
A coated steel and plastic tape dispenser with 2-inch capacity and an optional weighted base that is designed to hold Micronova 3-inch core tape rolls.
Have Questions?
Catalog NumberProduct TypeWidth (English)
MICP-52Tape Dispenser3.25 in.
Catalog number MICP-52
Price (SGD)
-
Product Type:
Tape Dispenser
Width (English):
3.25 in.
Micronova Tape Dispensers, For Use With: Film-backed or Paper tapes, Quantity: Each, Recommended Applications: Safety, Width: 2 in.
The Micronova Tape Dispenser is a combination of coated steel and plastic with a 2-inch width. Available with or without a weighted base. Designed for use with all Micronova 3-inch core tapes.
  • Coated steel and plastic tape dispenser
  • 2-inch capacity
  • Holds tapes with 3 inch cores
  • Designed for use with Micronova tapes
  • Optional weighted base
Specifications
Product TypeTape Dispenser
ColorBeige
Width (English)3.25 in.
MaterialPlastic
For Use With (Application)Film-backed or Paper tapes