Hand Held Scrubbing Tool
Hand Held Scrubbing Tool
Actual product may vary
Micronova
Hand Held Scrubbing Tool
A hand-held scrubbing tool that conforms to the hand and works with a choice of mop covers to ergonomically clean and scrub surfaces for a complete sanitization of small equipment and production surfaces.
Have Questions?
Catalog NumberMaterialProduct TypeQuantity
MICHHT-06PPPolyethyleneMop1 Ea.
Catalog number MICHHT-06PP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Material:
Polyethylene
Product Type:
Mop
Quantity:
1 Ea.
The Micronova Hand Held Mop is available in polypropylene, recyclable plastic or an autoclavable polyketone. Designed for use with se with MZSM 6-inch scrubbing pads or sanitizing covers to meet a variety of cleaning needs.
  • Hand-held cleaning and scrubbing mop with cover
  • Ideal for cleaning small equipment and production surfaces
  • Ergonomic, lightweight and conforms to hand
  • Available in Polypropylene, recyclable plastic or autoclavable Polyketone
  • Slip-on covers for specialized cleaning
    • NovaPoly™ Cover - 100% smooth knit polyester, matches traditional cleanroom wipe fabrics and provides excellent disinfectant dispersal
    • PolyMesh™ Cover - 100% polyester mesh, with foam interior, for scrubbing
  • Compatible with alcohol and strong disinfectants
  • Launderable for reuse
  • Autoclavable
Specifications
Dimensions15.2 x 10.2 cm (6 x 4 in.)
Product TypeMop
ColorWhite
Length (English)6 in.
SterilityNon-sterile
Length (Metric)15.2 cm
AutoclavableNot Autoclavable
Width (Metric)10.2 cm
Width (English)4 in.
Resistant ToChemicals
DisposableReusable
Quantity1 Ea.
Care InstructionsStore at Ambient Temperature
For Use With (Equipment)MZSM6-6NS or MZSM68-6S
DescriptionHand Held
For Use With (Application)Cleaning Small Equipment and Work Surfaces in Lab or Production
Certifications/ComplianceCertificate of Conformance upon Request
MaterialPolyethylene
Unit SizeEA