Surface Treated Sterile Tissue Culture Flasks, Plug Seal Cap
Actual product may vary
Surface Treated Sterile Tissue Culture Flasks, Plug Seal Cap
Perfect for cell growth.
Have Questions?
Catalog NumberCapacity (Metric)Culture Area
LSVFB012938250 mL75 cm2
Catalog number LSVFB012938
Price (SGD)
155.00
Case
Capacity (Metric):
250 mL
Culture Area:
75 cm2
Price (SGD)
155.00
Case
  • Flat, free flask surface
  • Striation free to maximize usable growth area
  • Used in both open, closed systems
  • Angled neck design offers good pipet and cell scraper access
  • Upper triangular, wider base shape provides stability
  • Protruding ridge on back side of flask makes it easy for stacking
  • Special area near neck for easy mark writing
  • Both flask sides have engraved graduation
  • Sterilized by gamma irradiation
  • Non-pyrogenic
Specifications
Certifications/ComplianceCE
Closure ColorBlue
ColorNatural
Capacity (Metric)250 mL
For Use With (Application)Cell growth
Neck StyleAngled
No. per Case100 Flasks
No. per Pack5 Flasks
Product TypeTissue Culture Flask
Surface CoatingSurface-treated
Writing SurfaceFlat
GraduatedYes
SterilitySterile
Bottom ShapeFlat
Closure TypePlug Seal
Culture Area75 cm2
Unit SizeCase