Serological Pipet 2ml, Sterilized, Non-P
Product Image
Serological Pipet 2ml, Sterilized, Non-P
Have Questions?
Catalog number LSV14955154
Price (SGD)
58.00
Special offer
Ends: 31-Mar-2026
76.00
Save 18.00 (24%)
Case
Add to cart
Price (SGD)
58.00
Special offer
Ends: 31-Mar-2026
76.00
Save 18.00 (24%)