Rotary Vane Vacuum Pumps
Rotary Vane Vacuum Pumps
Actual product may vary
Rotary Vane Vacuum Pumps
Designed for use with Labconco Freeze-dry Systems—compact size easily fits into freeze-dry system cabinets
Have Questions?
Change viewbuttonViewtableView
Catalog NumberAmperageFlow RatePlug Type
LBC77394022.4 A117 L/min.Reverse IEC
FIS16-315-512.4 A117 L/min.NEMA 6-15P
Catalog number LBC7739402
Price (SGD)
8,893.00
EA
Add to cart
Amperage:
2.4 A
Flow Rate:
117 L/min.
Plug Type:
Reverse IEC
Price (SGD)
8,893.00
EA
Add to cart
Designed for use with Labconco Freeze-dry Systems—compact size easily fits into freeze-dry system cabinets
Provide a reliable vacuum source for other products including glove boxes and centrifugal concentrators.
  • Two-stage, oil-sealed pumps with sliding vane
  • Switchable high-vacuum and high-throughput modes
  • Three-position gas ballast (Closed, Low Flow, and High Flow)
  • Enclosed, fan-cooled, single-phase electric motor drives pump
  • Thermal overload protection with automatic restart
  • Inlet valve prevents air and oil suckback in the event of power or oil loss
  • Cast-aluminum housing
  • Mounted on a base plate with rubber feet for stability
Cat. No. 16-108-218 has a reverse IEC plug; models with 117L/min. pumping capacity include a retractable lifting handle; models with 195L/min. pumping capacity have a lifting bracket. A pump exhaust filter with oil and odor filter elements is also included.
Specifications
Max. Vacuum2 x 10-3 mBar
Amperage2.4 A
IncludesPump, four each one liter bottles of vacuum pump oil and one exhaust filter with oil mist and odor filter elements. ON/OFF switch, power cord with plug for direct connection of pump to Labconco freeze dryers, one 12.7mm (1/2 in.) O.D. and one 19.1mm (3/4 in.) O.D. inlet adapter
Electrical Requirements230 V 50/60 Hz
MaterialCast Aluminum
Plug TypeReverse IEC
Weight (English)70 lb.
Weight (Metric)32 kg
Flow Rate117 L/min.
For Use WithFreeze dry system
Frequency50/60 Hz
Overall Height (English)10.4 in.
Overall Height (Metric)26.4 cm
Overall Length (English)17 in.
Overall Length (Metric)43.18 cm
Overall Width (English)6.5 in.
Pump TypeRotary Vane Vacuum Pump
Maximum Pressure1 atm