A5 Sterile Over Boots with Anti-slip Sole
Actual product may vary
Ansell
A5 Sterile Over Boots with Anti-slip Sole
Sterile cleanroom over boots with anti-slip soles featuring innovative Clean-Don™ technology for aseptic donning. Suitable for EU GMP ISO 5 Grade A sterile cleanrooms, the over boots are vacuum-packed and made from light and breathable fabric.
Have Questions?
Catalog NumberSize
KLC88808Universal
Catalog number KLC88808
Price (SGD)
840.00
Special offer
Ends: 06-Sep-2026
1,012.00
Save 172.00 (17%)
Case
Size:
Universal
Price (SGD)
840.00
Special offer
Ends: 06-Sep-2026
1,012.00
Save 172.00 (17%)
Case
  • Assured Compliance:
    • PPE Cat I according to Regulation (EU) 2016/425.
    • Certificate of Conformance by lot available online.
    • Certificate of Sterility by lot available online.
  • Suitable for use in EU GMP ISO 5 Class A sterile cleanroom environments and laboratories, and provided with supporting technical information to demonstrate regulatory compliance.
  • Featuring advanced Clean-Don™ technology for fast, simple and compliant aseptic donning, and anti-slip soles.
  • The comfortable over boots are provided in a small or a universal size and are made from low-lint breathable SMS fabric with a cloth-like feel.
  • Contamination protection is ensured through high quality sterility assured material with high-strength bound and triple-stitched seams.
  • Pair vacuum-packed and efficiently double-bagged to save space.
RightCycle
Specifications
SizeUniversal
MaterialSMS
Latex FreeYes
ColorWhite
DisposableSingle-use
SterilitySterile
Closure TypeElastic Top with Ankle Ties
Sole TypeAnti-slip
Packaging Quantity100 Pairs/Cs.
Packaging TypeTriple Bagged, Vacuum Packed
ResistanceNone
Product TypeControlled Environments Boot Covers
Cleanroom ClassClass 100 (ISO 5)
Unit SizeCase