Search Fisher Scientific
| Catalog Number | Size |
|---|---|
| KLC88802 | Large |
| FIS19-057-475 | Small |
| KLC88800 | Small |
| FIS19-057-476 | Medium |
| KLC88801 | Medium |
| FIS19-057-490 | Large |
| FIS19-057-491 | X-Large |
| KLC88803 | X-Large |
| FIS19-057-492 | 2X-Large |
Kimtech™ A5 Sterile Cleanroom Coveralls have an average bacterial filtration efficiency (BFE) of 96% and are made of 100% polypropylene breathable SMS fabric, which gives the wearer cloth-like comfort.
The Reflex Coverall Design is 7.5 times more rip-resistant than ANSI minimum standards and offers excellent fit and freedom of movement for the wearer. This personal protective apparel is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms.
It is not made with natural rubber latex, silicone, or the BHT preservative and is designed for single-use (disposable PPE). The easy-to-don, individually packaged coveralls feature a mandarin collar, thumb loops and tunneled elastics.