Microscope Slide Mailer
Actual product may vary
Microscope Slide Mailer
Allows for the safe storage and transport of microscope slides.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSlide Capacity
KLC621022
KLC621035
Catalog number KLC62102
Price (SGD)
23.00
Special offer
Ends: 31-Dec-2026
282.00
Save 259.00 (92%)
Box
Slide Capacity:
2
Price (SGD)
23.00
Special offer
Ends: 31-Dec-2026
282.00
Save 259.00 (92%)
Box
  • These white polypropylene slide mailers included hinged covers and molded-in dividers to securely hold 3 x 1 in. microscope slides.
  • Available in four sizes.
  • These white polypropylene slide mailers included hinged covers and molded-in dividers to securely hold 3 x 1 in. microscope slides.
  • Available in four sizes.
Specifications
TypeSlide Mailer
Slide Capacity2
For Use With (Equipment)Microscope Slides
Unit SizeBox