Kimtech™ Sterling™ 9.5 in. Nitrile Gloves
Kimtech™ Sterling™ 9.5 in. Nitrile Gloves
Actual product may vary
Kimtech™ Sterling™ 9.5 in. Nitrile Gloves
Sterling™ gloves combine the sensitivity of latex with the protection and strength of nitrile.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC50705X-Small
FIS19-048-132Small
KLC50706Small
KLC50707Medium
KLC50708Large
Catalog number KLC50705
Price (SGD)
531.00
CS
Add to cart
Size:
X-Small
Price (SGD)
531.00
CS
Add to cart
Sterling™ gloves combine the sensitivity of latex with the protection and strength of nitrile.
  • Standard 9.5 in. length
  • Powder free
  • Non-sterile
  • Static dissipative
  • Textured fingertips
  • Beaded cuff
  • Ambidextrous
  • Typical uses include: Healthcare, Laboratory, Cleaning, Maintenance, Electronic Assembly and Inspection
Specifications
Certifications/ComplianceASTM D 6319, Meets USP <800> guidelines
Certifications/Compliance (Europe)CE
Chemotherapeutic RatedNo
ApplicationsHealthcare, Laboratory
AQL1
ColorGray
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)3.5 mil
Finger Thickness (Metric)0.08 mm
HandSpecific
Latex FreeYes
MaterialNitrile
Palm Thickness (English)3.1 mil
Palm Thickness (Metric)0.07 mm
PowderedPowder Free
SizeX-Small
Thickness (English)3.5 mil
Thickness (Metric)0.08 mm
FDA 510 (k)Yes
For Use With (Application)Permeation resistance to most acids, aldehydes, alcohols, amides, bases, esters and ketones
Glove Cuff StyleGauntlet/Rolled
Grip DesignSmooth
LiningUnlined
Manufacturing/QMS Audit StandardsISO 9001
Model50705
SeriesKimtech™
Tensile Strength42 MPa
Length (English)9.5 in.
Length (Metric)24.13 cm
Unit SizeCS