Kimtech™ A7 Ankle High Shoe Covers
Actual product may vary
Kimtech™ A7 Ankle High Shoe Covers
Ankle high shoe covers provide extra contamination protection by covering the shoelace area with two elastic bands--one above the ankle and one below the ankle.
Have Questions?
Catalog NumberSize
KLC47972Universal
Catalog number KLC47972
Price (SGD)
284.00
Case
Size:
Universal
Price (SGD)
284.00
Case
  • Ankle high shoe covers provide 30% more protection that a shoe cover, but less than a boot
  • Two elastic bands placed both above and below the ankle for enhanced fit and coverage
  • Comfortably cover the shoelace area, closing a gap that could be a vehicle for contamination
  • Deliver a higher barrier of liquid protection and a securer grip with anti-skid vinyl soles
  • Kimtech A7 material offers enhanced chemical and biological protection (passes ASTM F1670 for penetration of blood and body fluids)
  • Recyclable through the RightCycle Program, which turns them into the raw materials to create eco-friendly consumer goods
  • Packaged 300 Shoe Covers/Case, 3 Bags of 100 Covers
Specifications
SizeUniversal
MaterialSMS
Latex FreeYes
ColorWhite
DisposableSingle-use
SterilityNon-sterile
Closure TypeOver and Under Elastic Ankle Bands
Sole TypeAnti-skid
Certifications/ComplianceASTM F1670
Packaging Quantity300/Cs.
Packaging Type3 Bags of 100 Covers
Product TypeControlled Environments Ankle High Shoe Covers
Unit SizeCase