Kimtech™ A6 Liquid Splash Protection Hooded Coveralls
Actual product may vary
Kimtech™ A6 Liquid Splash Protection Hooded Coveralls
Cleanroom hooded coveralls provide liquid barrier protection with a breathable back panel for use in ISO 7 and 8 environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC47681Small
FIS19-169-223BMedium
KLC47682Medium
FIS19-169-223CLarge
KLC47683Large
KLC47684X-Large
Catalog number KLC47681
Price (SGD)
-
Size:
Small
  • Presented unzipped with the inside of the garment first
  • Arms telescope folded
  • Legs telescope folded with built-in snaps
  • Elastic cuffs and thumb loops
  • No label on the outer surface
  • Hood, front panel, arms and leg made of high performance laminated SMS fabric
  • Back panel made of ultralight weight breathable SMS
  • 1 large bag of 25 coveralls individually packed per case
Specifications
Product TypeCleanroom Coveralls
SizeSmall
MaterialSMS
DisposableSingle-use
Cuff StyleElastic
Neck StyleHooded
SterilityNon-sterile
Certifications/CompliancePPE Cat III directive 89/686/EEC, Type 6 limited chemical splash protection; Type 5 particle protection; EN 14126:2003 certified protection against infective agent penetration
For Use With (Application)Controlled Environments
Cleanroom ClassClass 10,000-100,000 (ISO 7-8)
Unit SizeCase