White Anti-skid Shoe Cover with Black Vinyl Sole
Actual product may vary
White Anti-skid Shoe Cover with Black Vinyl Sole
Anti-skid shoe covers use proprietary fabric that helps filter out hazardous particulates as small as 0.3 microns in the 95 to 99% range and resist abrasions and use breathable fabric to keep the feet cool.
Have Questions?
Catalog number KLC4100
Price (SGD)
-
  • Shoe covers resist abrasions and use breathable fabric to reduce heat buildup
  • Secure elastic closures help keep shoe covers in place, while an anti-skid sole provides better traction to avoid slips, trips, and falls
  • Vinyl soles help improve traction to reduce slips, trips, and falls
  • A high moisture vapor transmission rate (MVTR) allows heat and moisture vapor to escape so wearers stay cooler
Specifications
SizeUniversal
MaterialMicroporous Laminate
Latex FreeYes
CoatingNone
ColorWhite
DisposableSingle-use
SterilityNon-sterile
Closure TypeElastic Top
Sole TypeAnti-skid
ResistanceAirborne Solid Particulates, Dry Particulates, Abrasion, Liquid Aerosol
Product TypeShoe Covers
Unit SizeCase