KleenGuard™ Unispec II Safety Eyewear
Actual product may vary
KleenGuard™ Unispec II Safety Eyewear
KleenGuard™ Unispec II Safety Eyewear offers wraparound protection to fit over most prescription eyewear
Have Questions?
Catalog number KLC16727
Price (SGD)
-
  • Metal-free design with screw-less hinges
  • Clear glasses (with clear temples) allow maximum visible light transmission; ideal for indoor use
  • Hard coated lenses for scratch resistance and longer life
  • Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
  • Meet the ANSI Z87.1+ standards for impact and are TAA compliant
Specifications
Lens CoatingHardcoat
Certifications/ComplianceMeets ANSI Z87.1+ standards for high impact (16727-01 and 16727-02 meet ANSI Z87.1+2010 and 16727-03 meets ANSI Z87.1+2015) and certified to meet the requirements of CSA Z94.3
Lens TintClear
No. per Pack50/Pk.
Unit SizeBox