KingFisher™ Plastics for 96 deep-well format
KingFisher™ Plastics for 96 deep-well format
Actual product may vary
KingFisher™ Plastics for 96 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
KFK97003530Combi PackKingFisher Duo Prime only
FKS1037348096 Deep Well plate, v-bottomKF Duo Prime, Flex and Presto
FKS11744978Tip CombKingFisher Flex and Presto
Catalog number KFK97003530
Price (SGD)
-
Product Type:
Combi Pack
For Use With (Equipment):
KingFisher Duo Prime only
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Product TypeCombi Pack
Format96 Deep-well Plate
DescriptionKingFisher combi pack for microtiter 96 deep-well plate (includes 8 plates, 8 tip combs, and 8 elution strips and caps) (for Duo Prime only).
For Use With (Equipment)KingFisher Duo Prime only
Unit SizeEach