Sure-Grip™ EX Flammable Safety Cabinet
Sure-Grip™ EX Flammable Safety Cabinet
Actual product may vary
Sure-Grip™ EX Flammable Safety Cabinet
When using a Sure-Grip EX Flammable Safety Cabinets, these two-door and bifold door cabinets protect workers, reduce fire risks, and improve productivity by allowing storage of flammable fuels and chemicals in code-compliant, sturdy 18-gauge steel structures.
Have Questions?
Catalog NumberColorLoad Bearing CapacityDoor StyleNo. of Doors
JUS894520Yellow45 gal.Self-close2
Catalog number JUS894520
Price (SGD)
-
Color:
Yellow
Load Bearing Capacity:
45 gal.
Door Style:
Self-close
No. of Doors:
2
Features of Justrite Flammables Cabinets
  • Double-wall construction provides 1.5 in. of insulating air space to help prevent the spread of flames
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Lettering on door reads: FLAMMABLE—KEEP FIRE AWAY
  • Accepts padlock (not included)
  • Color: yellow, gray (optional), white (19-160-376)
Specifications
Certifications/ComplianceFM, NFPA, OSHA
Depth (English) Interior14.56 in.
Depth (Metric) Exterior45.7 cm
Depth (Metric) Interior37 cm
Dimensions (D x W x H) Interior14.56 x 39.5 x 60.5 in.
Door StyleSelf-close
For Use With (Application)Flammable liquid storage and protection
Height (English) Interior60.5 in.
Height (Metric) Exterior165.1 cm
Height (Metric) Interior153.7 cm
IncludesU-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with
Load Bearing Capacity45 gal.
No. of Doors2
No. of Shelves2
Surface CoatingEpoxy, Polyester
Weight (English)392 lb.
Weight (Metric)178 kg
Width (English) Interior39.5 in.
Width (Metric) Exterior109 cm
Width (Metric) Interior100.3 cm
ColorYellow
Depth (English) Exterior18 in.
Height (English) Exterior65 in.
MaterialSteel (CR)
Product TypeCan Storage Cabinet
Width (English) Exterior43 in.
Unit SizeEA