SUB Aqua Pro Water Bath
Actual product may vary
SUB Aqua Pro Water Bath
SUB Aqua Pro Water Bath
Grant
SUB Aqua Pro Water Bath
Includes non-drip clear lid for improved performance, reduced evaporation and increased energy conservation. Grant Instruments™ SUB Aqua Pro Water Bath provides clear, wide-angle viewing LED display for instant reassurance of bath status.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Model
GRASAP3434 LSAP34
GRASAP1212 LSAP12
GRASAP1818 LSAP18
GRASAP22 LSAP2
GRASAP2626 LSAP26
FKS151470255 LSAP5
GRASAP55 LSAP5
GRASAPD5 and 12 LSAPD
GRASAP2S2 LSAP2s
Catalog number GRASAP34
Price (SGD)
6,530.00
Each
Capacity (Metric):
34 L
Model:
SAP34
Price (SGD)
6,530.00
Each
Includes non-drip clear lid for improved performance, reduced evaporation and increased energy conservation. Grant Instruments™ SUB Aqua Pro Water Bath provides clear, wide-angle viewing LED display for instant reassurance of bath status.
  • Ambient +5 to 99°C operation
  • Set and Forget™ technology - fast heat-up, accurate temperature control
  • Stability ±0.2°C
  • 0.1°C setting resolution
  • Adjustable over temperature alarm - protect samples from over heating
  • Advanced dry start and run dry protection
  • Suitable for use with heat transfer beads (excluding SAP2 & SAP2S models)
  • Drain tap and transparent lid included
  • Timer and presets
  • 120V & 230V models available
  • 3 year warranty
Specifications
Capacity (Metric)34 L
Temperature RangeAmbient +5° to 99°C
Heating Capacity1800 w (230V), 1300 w (120V)
ModelSAP34
Temperature Presets3
Temperature Resolution±0.1°C
Temperature Stability±0.2°C
Warranty3 Years
Dimensions (L x W x H)770 x 335 x 275 mm
Length (Metric)770 mm
Length (Metric) Interior635 mm
Width (Metric)335 mm
Width (Metric) Interior281 mm
Height (Metric)275 mm
Weight (Metric)13.8 kg
Voltage120/230 V