Polycarbonate Transparent Lid, Gabled
Polycarbonate Transparent Lid, Gabled
Actual product may vary
Polycarbonate Transparent Lid, Gabled
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
GRAAQL12Model SAP12, JBN12, JBA12 and SBB Aqua 12 Plus SUB Aqua Plus water bath
FKS11496417Model SAP12, JBN12, JBA12 and SBB Aqua 12 Plus SUB Aqua Plus water bath
FKS114864175L Tanks. Fits the Grant JBA5 bath, and would be a valid replacement lid for the JBN5.
Catalog number GRAAQL12
Price (SGD)
446.00
EA
Add to cart
For Use With (Equipment):
Model SAP12, JBN12, JBA12 and SBB Aqua 12 Plus SUB Aqua Plus water bath
Price (SGD)
446.00
EA
Add to cart
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
  • Polycarbonate construction
  • Clear, gabled lid allows condensation buildup to drip away from immersed vessels.
  • For use with Sub Aqua Pro Series Unstirred Water Baths
Specifications
DescriptionGabled
For Use With (Equipment)Model SAP12, JBN12, JBA12 and SBB Aqua 12 Plus SUB Aqua Plus water bath
For Use With (Application)Used to avoid contamination, reduce evaporation, and save energy
MaterialPolycarbonate
Unit SizeEA