Check Order Status
Zinc Sulfate Heptahydrate, Certified AR for Analysis, 99.5-100.5%, Fisher Chemical™
Element Search
Structure Search
Zinc Sulfate Heptahydrate, Certified AR for Analysis, 99.5-100.5%, Fisher Chemical™
H14O11SZn, CAS Number-7446-20-0, zinc sulfate heptahydrate, znso4.7h2o, zinc sulfate jan, zinc sulfate znso4 heptahydrate, white vitriol heptahydrate, ccris 5563, zinc sulfate heptahydrate 1:1:7, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, 500g, CHEBI:32312, White
Have Questions?
Catalog Number
Quantity
Packaging
GEEZ160053
also known as GEEZ/1600/53
500 g
Plastic powder jar
Catalog number
GEEZ160053
also known as GEEZ/1600/53
Price (SGD)
89.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Price (SGD)
89.00
EA
Add to cart
Specifications
Melting Point
100°C
CAS
7446-20-0
Color
White
pH
4.4 to 6
Physical Form
Solid
View more
H14O11SZn, CAS Number-7446-20-0, zinc sulfate heptahydrate, znso4.7h2o, zinc sulfate jan, zinc sulfate znso4 heptahydrate, white vitriol heptahydrate, ccris 5563, zinc sulfate heptahydrate 1:1:7, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, 500g, CHEBI:32312, White