▸
Water and Wastewater Testing Supplies
▸
Water Quality Testing Kits
▸
Platinum Cobalt Color Standard Solution (APHA), Fisher Chemical™
Actual product may vary
Fisher Chemical
Platinum Cobalt Color Standard Solution (APHA), Fisher Chemical™
Have Questions?
Change view
Catalog Number
Quantity
GEEUSSP120500
500 mL
FISSP120-500
500 mL
GEEUSSP120-500
500 mL
3 Options
Catalog number
GEEUSSP120500
Price (SGD)
420.00
Each
Add to cart
Quantity:
500 mL
Price (SGD)
420.00
Each
Add to cart
Specifications
Color
Yellow
pH Range
1.1
Format
Liquid
Packaging
Glass Bottle
Quantity
500 mL
Type
Platinum Cobalt Color Standard Solution