Check Order Status
Sodium Sulfate Anhydrous, 99+%, Extra Pure, Fisher Chemical
Element Search
Structure Search
Sodium Sulfate Anhydrous, 99+%, Extra Pure, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
GEES660060
also known as GEES/6600/60
1 kg
Plastic powder jar
Catalog number
GEES660060
also known as GEES/6600/60
Price (SGD)
35.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Price (SGD)
35.00
EA
Add to cart
Specifications
Melting Point
884°C
CAS
7757-82-6
Color
White
pH
5 to 8
Physical Form
Solid
View more