Check Order Status
Sodium Sulfate Anhydrous, 99+%, Extra Pure, Fisher Chemical™
Element Search
Structure Search
Sodium Sulfate Anhydrous, 99+%, Extra Pure, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
GEES660053
also known as GEES/6600/53
500 g
Catalog number
GEES660053
also known as GEES/6600/53
Price (SGD)
30.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
30.00
EA
Add to cart
Specifications
Melting Point
884°C
CAS
7757-82-6
Color
White
pH
5 to 8
Physical Form
Solid
View more
Sodium Sulfate Anhydrous, 99+%, Extra Pure