Check Order Status
Sodium Carbonate Anhydrous, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Sodium Carbonate Anhydrous, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
GEES292053
also known as GEES/2920/53
500 g
Plastic powder jar
Catalog number
GEES292053
also known as GEES/2920/53
Price (SGD)
78.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Price (SGD)
78.00
EA
Add to cart
Specifications
Boiling Point
1600°C
Melting Point
854°C
CAS
497-19-8
Color
White
pH
11.3
View more
Sodium Carbonate Anhydrous, Certified AR for Analysis