Silver Nitrate, Certified AR for Analysis, Fisher Chemical™
Silver Nitrate, Certified AR for Analysis, Fisher Chemical™
Silver Nitrate, Certified AR for Analysis, Fisher Chemical™
Silver Nitrate, Certified AR for Analysis, Fisher Chemical™
AgNO3, CAS Number-7761-88-8, silbernitrat, silver i nitrate, silver mononitrate, nitrate d'argent, silver nitrate, nitric acid silver i salt, silvernitrate, argenti nitras, argerol, lunar caustic, 500g, 444 deg.C, CHEBI:32130, White, 169.87g/mol, SQGYOTSLMSWVJD-UHFFFAOYSA-N, silver;nitrate
Have Questions?
Catalog NumberQuantity
GEES128048
also known as GEES/1280/48
100 g
Catalog number GEES128048
also known as GEES/1280/48
Price (SGD)
453.00
Each
Quantity:
100 g
Request bulk or custom format
Price (SGD)
453.00
Each
Specifications
Boiling Point444°C
Melting Point212°C
CAS7761-88-8
ColorWhite
pH5.4 to 6.4
View more
AgNO3, CAS Number-7761-88-8, silbernitrat, silver i nitrate, silver mononitrate, nitrate d'argent, silver nitrate, nitric acid silver i salt, silvernitrate, argenti nitras, argerol, lunar caustic, 500g, 444 deg.C, CHEBI:32130, White, 169.87g/mol, SQGYOTSLMSWVJD-UHFFFAOYSA-N, silver;nitrate