Check Order Status
Isopropanol, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Isopropanol, Certified AR for Analysis, Fisher Chemical™
C3H8O, CAS Number-67-63-0, 2-hydroxypropane, isopropanol, hartosol, 2-propanol, dimethylcarbinol, avantine, petrohol, sec-propyl alcohol, isopropyl alcohol, alkolave, 2.5L, 81 deg.C, CHEBI:17824, Colorless, 60.1g/mol, KFZMGEQAYNKOFK-UHFFFAOYSA-N, propan-2-ol, -89.5 deg.C, 11674, 60.096 | CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
GEEP750021
also known as GEEP/7500/21
5 L
HDPE Bottle
Catalog number
GEEP750021
also known as GEEP/7500/21
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 L
Packaging:
HDPE Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Isopropanol
pH
7
Melting Point
-89.5°C
CAS
67-63-0
Color
Colorless
View more
C3H8O, CAS Number-67-63-0, 2-hydroxypropane, isopropanol, hartosol, 2-propanol, dimethylcarbinol, avantine, petrohol, sec-propyl alcohol, isopropyl alcohol, alkolave, 2.5L, 81 deg.C, CHEBI:17824, Colorless, 60.1g/mol, KFZMGEQAYNKOFK-UHFFFAOYSA-N, propan-2-ol, -89.5 deg.C, 11674, 60.096