Check Order Status
Magnesium Chloride Hexahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Element Search
Structure Search
Magnesium Chloride Hexahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
GEEM060053
also known as GEEM/0600/53
500 g
Plastic powder jar
Catalog number
GEEM060053
also known as GEEM/0600/53
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
Melting Point
117°C
CAS
7791-18-6
Color
White
pH
5 to 6.5
Physical Form
Powder/Solid
View more