Check Order Status
Karl Fischer Aqualine™ Water Standard 1.0mg, for Karl Fischer Titration, Fisher Chemical
Element Search
Structure Search
Karl Fischer Aqualine™ Water Standard 1.0mg, for Karl Fischer Titration, Fisher Chemical
1mg/mL H
2
O standard
Have Questions?
Catalog Number
Quantity
GEEK271099
also known as GEEK/2710/99
10 x 4 mL
Catalog number
GEEK271099
also known as GEEK/2710/99
Price (SGD)
232.00
EA
Add to cart
Quantity:
10 x 4 mL
Request bulk or custom format
Price (SGD)
232.00
EA
Add to cart
Specifications
Chemical Name or Material
Aqualine™ Standard 1.1
Color
Colorless
Quantity
10 x 4 mL
Cleanup Symbol
Solvent
Name Note
Karl Fischer Reagent
View more
Standard contains 1mg H
2
O/mL (1000ppm)
Used for titer determination of coulometric or volumetric Karl Fischer reagents
Supplied as 10 x 4mL and packaged in glass ampules to protect from moisture
The ampule should only be opened immediately prior to use to prevent absorption of moisture from atmosphere