Check Order Status
Karl Fischer Aqualine™ Matrix K, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Element Search
Structure Search
Karl Fischer Aqualine™ Matrix K, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
GEEK2300R15
also known as GEEK/2300R/15
1 L
Catalog number
GEEK2300R15
also known as GEEK/2300R/15
Price (SGD)
76.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
76.00
EA
Add to cart
Chemical Identifiers
CAS
107-21-1, 111-76-2, 5683-33-0
Specifications
Chemical Name or Material
Aqualine™ Matrix K
Color
Colorless
Quantity
1 L
Cleanup Symbol
Solvent
Name Note
Karl Fischer Reagent
View more
Matrix to be used in conjunction with Complete 5K (US: AL2250R; EU: K/2250R) for high water content analysis of aldehydes and ketones.