Check Order Status
Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Element Search
Structure Search
Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
GEEJ433015
also known as GEEJ/4330/15
1 L
HDPE plastic bottle
Catalog number
GEEJ433015
also known as GEEJ/4330/15
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Color
Colorless
CAS
7647-01-0
Boiling Point
100°C
Packaging
HDPE plastic bottle
Quantity
1 L
View more
Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis