▸
Buffers and Standards
▸
General Chemistry Solutions
▸
Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
Hydrogen Peroxide, 30% (Certified ACS), Thermo Scientific™
Have Questions?
Catalog Number
Quantity
Packaging
GEEH325-500
500 mL
Poly Bottle
Catalog number
GEEH325-500
Price (SGD)
62.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
62.00
Each
Add to cart
Specifications
Chemical Name or Material
Hydrogen Peroxide, 30%
Color
Colorless
CAS
7722-84-1
ChemAlert Storage Symbol
Yellow
Boiling Point
108°C
View more
Hydrogen Peroxide, 30% (Certified ACS)