Hydrochloric Acid 37%, for Trace Metal Analysis, d=1.18, Primar Plus™, Fisher Chemical™
Hydrochloric Acid 37%, for Trace Metal Analysis, d=1.18, Primar Plus™, Fisher Chemical™
Hydrochloric Acid 37%, for Trace Metal Analysis, d=1.18, Primar Plus™, Fisher Chemical™
Hydrochloric Acid 37%, for Trace Metal Analysis, d=1.18, Primar Plus™, Fisher Chemical™
Hydrochloric Acid 37%, for Trace Metal Analysis, d=1.18, Primar Plus™, Fisher Chemical™
ClH, CAS Number-7647-01-0, anhydrous hydrochloric acid, acide chlorhydrique, chlorwasserstoff, hydrochloric acid, chloorwaterstof, chlorohydric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol
Have Questions?
Catalog NumberQuantity
GEEH1196PB15
also known as GEEH/1196/PB15
1 L
Catalog number GEEH1196PB15
also known as GEEH/1196/PB15
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
pH1
ColorColorless
CAS7647-01-0
Melting Point-35°C
Boiling Point57°C
View more
ClH, CAS Number-7647-01-0, anhydrous hydrochloric acid, acide chlorhydrique, chlorwasserstoff, hydrochloric acid, chloorwaterstof, chlorohydric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol