Check Order Status
N-Heptane, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
N-Heptane, Certified AR for Analysis, Fisher Chemical™
C7H16, CAS Number-142-82-5, dipropylmethane, dipropyl methane, eptani, n-heptane, gettysolve-c, heptan polish, skellysolve c, heptyl hydride, heptan, heptanen, 2.5L, 98 deg.C, CHEBI:43098, Colorless, 100.2g/mol, IMNFDUFMRHMDMM-UHFFFAOYSA-N, -91 deg.C, 9544, 100.205, Amber glass bottle, Liquid, 8900
Have Questions?
Catalog Number
Quantity
Packaging
GEEH016017
also known as GEEH/0160/17
2.5 L
Amber Glass Bottle
Catalog number
GEEH016017
also known as GEEH/0160/17
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Melting Point
-91°C
CAS
142-82-5
Color
Colorless
Boiling Point
98°C
Quantity
2.5 L
View more
C7H16, CAS Number-142-82-5, dipropylmethane, dipropyl methane, eptani, n-heptane, gettysolve-c, heptan polish, skellysolve c, heptyl hydride, heptan, heptanen, 2.5L, 98 deg.C, CHEBI:43098, Colorless, 100.2g/mol, IMNFDUFMRHMDMM-UHFFFAOYSA-N, -91 deg.C, 9544, 100.205, Amber glass bottle, Liquid, 8900