Check Order Status
Cyclohexane, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Cyclohexane, Certified AR for Analysis, Fisher Chemical™
C6H12, CAS Number-110-82-7, 2.5L, 81 deg.C, CHEBI:29005, Colorless, 84.15g/mol, XDTMQSROBMDMFD-UHFFFAOYSA-N, cyclohexane, 6.5 deg.C, 3814, 84.162, Amber glass bottle, Liquid, 8078, C1CCCCC1, 2.9, 104mbar at 20 deg.C, 0.94 mPaS at 20 deg.C
Have Questions?
Catalog Number
Quantity
Packaging
GEEC892117
also known as GEEC/8921/17
2.5 L
Amber Glass Bottle
Catalog number
GEEC892117
also known as GEEC/8921/17
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS
110-82-7
Specifications
Melting Point
6.5°C
CAS
110-82-7
Color
Colorless
Boiling Point
81°C
Quantity
2.5 L
View more
C6H12, CAS Number-110-82-7, 2.5L, 81 deg.C, CHEBI:29005, Colorless, 84.15g/mol, XDTMQSROBMDMFD-UHFFFAOYSA-N, cyclohexane, 6.5 deg.C, 3814, 84.162, Amber glass bottle, Liquid, 8078, C1CCCCC1, 2.9, 104mbar at 20 deg.C, 0.94 mPaS at 20 deg.C