Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™
Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™
Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™
Fisher Chemical
Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™
CAS: 7558-79-4 | Molecular Formula: HNa2O4P | Molecular Weight: 141.96 g/mol
Have Questions?
Catalog NumberQuantityPackaging
GEEBP332-500500 gGlass Bottle
Catalog number GEEBP332-500
Price (SGD)
84.00
Each
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
84.00
Each
Specifications
Chemical Name or MaterialSodium Phosphate Dibasic Anhydrous
CAS7558-79-4
ChemAlert Storage SymbolGray
ColorWhite
Health Hazard 1Signal Word: CAUTION!
View more
Sodium Phosphate Dibasic Anhydrous (White Granules or Powder)
This buffer component is commonly used in tissue culture and molecular biology applications.