D-(+)-Trehalose, Dihydrate, Fisher BioReagents
D-(+)-Trehalose, Dihydrate, Fisher BioReagents
D-(+)-Trehalose, Dihydrate, Fisher BioReagents
Fisher Chemical
D-(+)-Trehalose, Dihydrate, Fisher BioReagents
Have Questions?
Catalog NumberQuantityPackaging
GEEBP2687-100100 gPoly Bottle
Catalog number GEEBP2687-100
Price (SGD)
187.00
Each
Quantity:
100 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
187.00
Each
Specifications
Chemical Name or MaterialD-(+)-Trehalose, Dihydrate
CAS6138-23-4
ChemAlert Storage SymbolGray
Health Hazard 2WARNING!
Health Hazard 3Emergency Overview
Irritating to eyes and skin. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:2
Flammability:1
Instability:1
View more
D-(+)-Trehalose is used in several biochemical techniques.